siRNA Details
| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032462.2 | NP_115851.1 | 8 | 707 | ||
| XM_006234989.2 | XP_006235051.1 | 8 | 836 | |||
| XM_006234990.2 | XP_006235052.1 | 7 | 4152 | |||
| GenBank mRNA | AB043892.1 | - | 7 | 686 | ||
| AF297118.1 | - | 8 | 709 | |||

Kcnip3
Genomic Map