siRNA Details
| Gene Symbol: Slc16a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031744.2 | NP_113932.2 | 5 | 899 | ||
| XM_006242039.2 | XP_006242101.1 | 4 | 1168 | |||
| GenBank mRNA | AF059258.1 | - | 5 | 899 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a8| Gene Symbol: Slc16a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031744.2 | NP_113932.2 | 5 | 899 | ||
| XM_006242039.2 | XP_006242101.1 | 4 | 1168 | |||
| GenBank mRNA | AF059258.1 | - | 5 | 899 | ||