siRNA Details
| Gene Symbol: Slc13a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031746.1 | NP_113934.1 | 4, 5 | 633 | ||
| GenBank mRNA | AB001321.1 | - | 4, 5 | 636 | ||
| AF058714.1 | - | 4, 5 | 633 | |||
| U51153.1 | - | 4, 5 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc13a2| Gene Symbol: Slc13a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031746.1 | NP_113934.1 | 4, 5 | 633 | ||
| GenBank mRNA | AB001321.1 | - | 4, 5 | 636 | ||
| AF058714.1 | - | 4, 5 | 633 | |||
| U51153.1 | - | 4, 5 | 625 | |||