siRNA Details
| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031747.1 | NP_113935.1 | 2, 3 | 244 | ||
| GenBank mRNA | BC061809.1 | - | 2, 3 | 213 | 5598777 | |
| D14437.1 | - | 2, 3 | 244 | |||
| X71071.1 | - | 2, 3 | 209 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnn1| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031747.1 | NP_113935.1 | 2, 3 | 244 | ||
| GenBank mRNA | BC061809.1 | - | 2, 3 | 213 | 5598777 | |
| D14437.1 | - | 2, 3 | 244 | |||
| X71071.1 | - | 2, 3 | 209 | |||