siRNA Details
| Gene Symbol: Rhag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023022.2 | NP_075411.2 | 1 | 222 | ||
| GenBank mRNA | AB015194.1 | - | 1 | 195 | ||
| AF531097.1 | - | 1 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhag| Gene Symbol: Rhag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023022.2 | NP_075411.2 | 1 | 222 | ||
| GenBank mRNA | AB015194.1 | - | 1 | 195 | ||
| AF531097.1 | - | 1 | 222 | |||