siRNA Details
| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023023.1 | NP_075412.1 | 12 | 1709 | ||
| GenBank mRNA | AB029432.1 | - | 11 | 1583 | ||
| AJ131436.1 | - | 12 | 1709 | |||
| Gene Symbol: LOC100911610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239841.2 | XP_006239903.1 | 4 | 1279 | ||
| Gene Symbol: LOC103692570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764540.1 | XP_008762762.1 | 1820 | |||

Dpysl5
Genomic Map