siRNA Details
| Gene Symbol: Prkaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019142.2 | NP_062015.2 | 7 | 895 | ||
| GenBank mRNA | U40819.1 | - | 7 | 862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkaa1| Gene Symbol: Prkaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019142.2 | NP_062015.2 | 7 | 895 | ||
| GenBank mRNA | U40819.1 | - | 7 | 862 | ||