siRNA Details
| Gene Symbol: Myo1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023092.1 | NP_075580.2 | 18 | 2030 | ||
| GenBank mRNA | X74800.1 | - | 18 | 2030 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo1c| Gene Symbol: Myo1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023092.1 | NP_075580.2 | 18 | 2030 | ||
| GenBank mRNA | X74800.1 | - | 18 | 2030 | ||