siRNA Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023093.1 | NP_075581.1 | 7 | 981 | ||
| GenBank mRNA | BC061830.1 | - | 7 | 981 | 5599635 | |
| FQ214146.1 | - | 6 | 823 | |||
| FQ225416.1 | - | 2 | 296 | |||
| FQ225735.1 | - | 2 | 296 | |||
| FQ227531.1 | - | 2 | 296 | |||
| FQ227720.1 | - | 1 | 116 | |||
| J05266.1 | - | 7 | 902 | |||

Atp5a1
Genomic Map