siRNA Details
| Gene Symbol: Mgat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023095.1 | NP_075583.1 | 15 | 2036 | ||
| XM_008769499.1 | XP_008767721.1 | 11 | 1580 | |||
| GenBank mRNA | L14284.1 | - | 15 | 2036 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgat5| Gene Symbol: Mgat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023095.1 | NP_075583.1 | 15 | 2036 | ||
| XM_008769499.1 | XP_008767721.1 | 11 | 1580 | |||
| GenBank mRNA | L14284.1 | - | 15 | 2036 | ||