siRNA Details
| Gene Symbol: Nup62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023098.2 | NP_075586.1 | 2 | 1576 | ||
| XM_006229150.2 | XP_006229212.1 | 2 | 1775 | |||
| XM_006229151.2 | XP_006229213.1 | 2 | 2481 | |||
| GenBank mRNA | BC128709.1 | - | 1 | 1480 | 8363131 | |
| J04143.1 | - | 1 | 334 | |||
| S75997.1 | - | 513 | ||||
| X52583.1 | - | 1 | 2156 | |||

Nup62
Genomic Map