siRNA Details
| Gene Symbol: Vti1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023101.1 | NP_075589.1 | 3 | 204 | ||
| GenBank mRNA | AF262221.1 | - | 3 | 204 | ||
| AF262222.1 | - | 3 | 190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vti1a| Gene Symbol: Vti1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023101.1 | NP_075589.1 | 3 | 204 | ||
| GenBank mRNA | AF262221.1 | - | 3 | 204 | ||
| AF262222.1 | - | 3 | 190 | |||