siRNA Details
| Gene Symbol: Arhgef9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023957.1 | NP_076447.1 | 6 | 1284 | ||
| GenBank mRNA | AJ250425.1 | - | 6 | 1284 | ||
| AJ302676.1 | - | 5 | 1104 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef9| Gene Symbol: Arhgef9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023957.1 | NP_076447.1 | 6 | 1284 | ||
| GenBank mRNA | AJ250425.1 | - | 6 | 1284 | ||
| AJ302676.1 | - | 5 | 1104 | |||