siRNA Details
| Gene Symbol: Kcns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023966.1 | NP_076456.1 | 2 | 1023 | ||
| GenBank mRNA | AJ296090.1 | - | 2 | 1023 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcns2| Gene Symbol: Kcns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023966.1 | NP_076456.1 | 2 | 1023 | ||
| GenBank mRNA | AJ296090.1 | - | 2 | 1023 | ||