siRNA Details
| Gene Symbol: Npy2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023968.1 | NP_076458.1 | 1 | 678 | ||
| XM_006232534.2 | XP_006232596.1 | 2 | 1444 | |||
| XM_008761113.1 | XP_008759335.1 | 2 | 773 | |||
| GenBank mRNA | AF054870.1 | - | 1 | 118 | ||
| AY004257.1 | - | 1 | 678 | |||
More Information
Set Membership: |
|

Npy2r
Genomic Map