siRNA Details
| Gene Symbol: Arl6ip5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023972.3 | NP_076462.1 | 2 | 334 | ||
| GenBank mRNA | AF240182.1 | - | 2 | 334 | ||
| BC061548.1 | - | 346 | 6922366 | |||
| FQ213351.1 | - | 367 | ||||
| FQ213919.1 | - | 356 | ||||
| FQ216190.1 | - | 367 | ||||
| FQ216551.1 | - | 368 | ||||
| FQ225681.1 | - | 367 | ||||
| FQ228780.1 | - | 432 | ||||
| FQ232358.1 | - | 367 | ||||

Arl6ip5
Genomic Map