siRNA Details
| Gene Symbol: Synpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023974.1 | NP_076464.1 | 2 | 375 | ||
| XM_008770570.1 | XP_008768792.1 | 2 | 587 | |||
| GenBank mRNA | AF306459.1 | - | 2 | 375 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synpr| Gene Symbol: Synpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023974.1 | NP_076464.1 | 2 | 375 | ||
| XM_008770570.1 | XP_008768792.1 | 2 | 587 | |||
| GenBank mRNA | AF306459.1 | - | 2 | 375 | ||