siRNA Details
Gene Symbol: Scn9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133289.1 | NP_579823.1 | 19 | 3737 | ||
GenBank mRNA | AF000368.1 | - | 19 | 3737 | ||
U79568.1 | - | 19 | 3737 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scn9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133289.1 | NP_579823.1 | 19 | 3737 | ||
GenBank mRNA | AF000368.1 | - | 19 | 3737 | ||
U79568.1 | - | 19 | 3737 |