siRNA Details
| Gene Symbol: Scn9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133289.1 | NP_579823.1 | 12 | 1984 | ||
| GenBank mRNA | AF000368.1 | - | 12 | 1984 | ||
| U79568.1 | - | 12 | 1984 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn9a| Gene Symbol: Scn9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133289.1 | NP_579823.1 | 12 | 1984 | ||
| GenBank mRNA | AF000368.1 | - | 12 | 1984 | ||
| U79568.1 | - | 12 | 1984 | |||