siRNA Details
| Gene Symbol: Bcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031752.2 | NP_113940.1 | 5 | 583 | ||
| GenBank mRNA | AB035510.1 | - | 5 | 580 | ||
| BC072479.1 | - | 5 | 583 | 7104047 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcam| Gene Symbol: Bcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031752.2 | NP_113940.1 | 5 | 583 | ||
| GenBank mRNA | AB035510.1 | - | 5 | 580 | ||
| BC072479.1 | - | 5 | 583 | 7104047 | ||