siRNA Details
| Gene Symbol: Prx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023976.2 | NP_076466.2 | 7 | 2820 | ||
| GenBank mRNA | Z29649.2 | - | 7 | 2820 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prx| Gene Symbol: Prx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023976.2 | NP_076466.2 | 7 | 2820 | ||
| GenBank mRNA | Z29649.2 | - | 7 | 2820 | ||