siRNA Details
| Gene Symbol: Npffr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023980.1 | NP_076470.1 | 3 | 1090 | ||
| XM_006250787.2 | XP_006250849.1 | 4 | 1651 | |||
| GenBank mRNA | AF268900.1 | - | 3 | 1090 | ||
More Information
Set Membership: |
|

Npffr2
Genomic Map