siRNA Details
| Gene Symbol: Csf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023981.4 | NP_076471.3 | 3, 4 | 393 | ||
| XM_006233187.1 | XP_006233249.1 | 3, 4 | 264 | |||
| XM_008761426.1 | XP_008759648.1 | 3, 4 | 264 | |||
| XM_008761427.1 | XP_008759649.1 | 3, 4 | 264 | |||
| XM_008761428.1 | XP_008759650.1 | 3, 4 | 264 | |||
| XM_008761429.1 | XP_008759651.1 | 3, 4 | 264 | |||
| GenBank mRNA | AF514355.1 | - | 3, 4 | 233 | ||
| AF514356.1 | - | 3, 4 | 233 | |||
| AF514357.1 | - | 3, 4 | 243 | |||
| AF515736.1 | - | 3, 4 | 331 | |||
| BC074007.1 | - | 3, 4 | 393 | 7099454 | ||

Csf1
Genomic Map