siRNA Details
| Gene Symbol: Zfp354c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023988.1 | NP_076478.1 | 4 | 344 | ||
| XM_006246335.2 | XP_006246397.1 | 5 | 1140 | |||
| XM_006246337.2 | XP_006246399.1 | 6 | 1373 | |||
| XM_008767714.1 | XP_008765936.1 | 6 | 1253 | |||
| GenBank mRNA | AF321874.1 | - | 4 | 344 | ||

Zfp354c
Genomic Map