siRNA Details
| Gene Symbol: Senp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023989.1 | NP_076479.1 | 10 | 941 | ||
| XM_008768805.1 | XP_008767027.1 | 10 | 1072 | |||
| GenBank mRNA | AF260129.1 | - | 10 | 941 | ||
| FQ212134.1 | - | 10 | 1036 | |||
| FQ222500.1 | - | 9 | 884 | |||

Senp2
Genomic Map