siRNA Details
| Gene Symbol: Prkaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023991.1 | NP_076481.1 | 3 | 308 | ||
| XM_008763901.1 | XP_008762123.1 | 2 | 2037 | |||
| GenBank mRNA | U12149.1 | - | 3 | 493 | ||
| Z29486.1 | - | 3 | 308 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkaa2| Gene Symbol: Prkaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023991.1 | NP_076481.1 | 3 | 308 | ||
| XM_008763901.1 | XP_008762123.1 | 2 | 2037 | |||
| GenBank mRNA | U12149.1 | - | 3 | 493 | ||
| Z29486.1 | - | 3 | 308 | |||