siRNA Details
| Gene Symbol: Mrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024001.1 | NP_076491.1 | 9 | 1050 | ||
| XM_006253937.2 | XP_006253999.1 | 9 | 1168 | |||
| GenBank mRNA | AF288289.1 | - | 9 | 1050 | ||
| AF288290.1 | - | 5 | 501 | |||
| AF288291.1 | - | 5 | 503 | |||
| BC083554.1 | - | 9 | 1043 | |||
| BC098916.1 | - | 9 | 1025 | |||

Mrs2
Genomic Map