siRNA Details
| Gene Symbol: Secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024002.1 | NP_076492.1 | 6 | 849 | ||
| XM_008771506.1 | XP_008769728.1 | 6 | 896 | |||
| GenBank mRNA | AJ251245.1 | - | 6 | 849 | ||
| FQ227169.1 | - | 6 | 869 | |||
| FQ231169.1 | - | 6 | 871 | |||
| FQ232320.1 | - | 6 | 877 | |||
| FQ234993.1 | - | 6 | 890 | |||

Secisbp2
Genomic Map