siRNA Details
| Gene Symbol: Pigm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024144.1 | NP_077058.1 | 1 | 557 | ||
| GenBank mRNA | AB028126.1 | - | 1 | 557 | ||
| Gene Symbol: LOC103694910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773907.1 | XP_008772129.1 | 2126 | |||

Pigm
Genomic Map