siRNA Details
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024146.1 | NP_077060.1 | 6 | 898 | ||
XM_006253323.2 | XP_006253385.1 | 6 | 906 | |||
XM_006253324.2 | XP_006253386.1 | 7 | 1113 | |||
XM_006253325.2 | XP_006253387.1 | 5 | 639 | |||
XM_006253326.2 | XP_006253388.1 | 5 | 633 | |||
XM_006253327.1 | XP_006253389.1 | 6 | 920 | |||
GenBank mRNA | D12498.1 | - | 6 | 898 | ||
S54008.1 | - | 6 | 925 |
