siRNA Details
| Gene Symbol: Arf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024151.1 | NP_077065.1 | 1, 2 | 283 | ||
| GenBank mRNA | BC063167.1 | - | 1, 2 | 241 | 6921397 | |
| FQ213125.1 | - | 1, 2 | 266 | |||
| FQ213804.1 | - | 1, 2 | 280 | |||
| FQ234083.1 | - | 1, 2 | 241 | |||
| FQ234248.1 | - | 1, 2 | 353 | |||
| L12383.1 | - | 1, 2 | 283 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007479, BC115650, D87901, AK081686, AK168793, and others.

Arf4
Genomic Map