siRNA Details
| Gene Symbol: Fdxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024153.1 | NP_077067.1 | 5 | 527 | ||
| GenBank mRNA | BC088844.1 | - | 5 | 534 | 7308634 | |
| D63761.1 | - | 5 | 527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fdxr| Gene Symbol: Fdxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024153.1 | NP_077067.1 | 5 | 527 | ||
| GenBank mRNA | BC088844.1 | - | 5 | 534 | 7308634 | |
| D63761.1 | - | 5 | 527 | |||