siRNA Details
| Gene Symbol: Asic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024154.2 | NP_077068.1 | 6 | 1020 | ||
| XM_006257378.2 | XP_006257440.1 | 4 | 1345 | |||
| XM_006257381.2 | XP_006257443.1 | 6 | 1305 | |||
| XM_008765793.1 | XP_008764015.1 | 4 | 1349 | |||
| XM_008765794.1 | XP_008764016.1 | 6 | 1308 | |||
| XM_008765795.1 | XP_008764017.1 | 6 | 961 | |||
| XM_008765796.1 | XP_008764018.1 | 6 | 1305 | |||
| GenBank mRNA | AB049451.1 | - | 4 | 611 | ||
| AJ006519.1 | - | 4 | 1066 | |||
| AJ309926.1 | - | 4 | 1070 | |||

Asic1
Genomic Map