siRNA Details
| Gene Symbol: Anxa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024155.3 | NP_077069.3 | 12 | 974 | ||
| XM_006236841.2 | XP_006236903.1 | 13 | 1061 | |||
| XM_006236842.2 | XP_006236904.1 | 12 | 1597 | |||
| GenBank mRNA | BC085688.1 | - | 12 | 974 | 7191458 | |
| D38224.2 | - | 10 | 883 | |||

Anxa4
Genomic Map