siRNA Details
| Gene Symbol: Dck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024158.1 | NP_077072.1 | 3, 4 | 438 | ||
| GenBank mRNA | FQ212366.1 | - | 743 | |||
| L33894.1 | - | 3, 4 | 438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dck| Gene Symbol: Dck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024158.1 | NP_077072.1 | 3, 4 | 438 | ||
| GenBank mRNA | FQ212366.1 | - | 743 | |||
| L33894.1 | - | 3, 4 | 438 | |||