siRNA Details
| Gene Symbol: Epha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169137.2 | NP_001162608.1 | 15 | 2917 | ||
| NM_024367.1 | NP_077343.1 | 14 | 2581 | |||
| XM_006250818.2 | XP_006250880.1 | 14 | 3363 | |||
| XM_006250819.2 | XP_006250881.1 | 14 | 3360 | |||
| XM_008770059.1 | XP_008768281.1 | 15 | 3429 | |||
| GenBank mRNA | X78689.1 | - | 14 | 2648 | ||

Epha5
Genomic Map