siRNA Details
| Gene Symbol: Epha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169137.2 | NP_001162608.1 | 11 | 2308 | ||
| NM_024367.1 | NP_077343.1 | 10 | 1972 | |||
| XM_006250818.2 | XP_006250880.1 | 10 | 2754 | |||
| XM_006250819.2 | XP_006250881.1 | 10 | 2751 | |||
| XM_008770059.1 | XP_008768281.1 | 11 | 2820 | |||
| GenBank mRNA | X78689.1 | - | 10 | 2039 | ||

Epha5
Genomic Map