siRNA Details
| Gene Symbol: Mtpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024374.4 | NP_077350.1 | 3 | 378 | ||
| GenBank mRNA | AY951952.1 | - | 385 | |||
| BC088136.1 | - | 3 | 378 | 7377576 | ||
| D26179.1 | - | 3 | 433 | |||
| FQ228349.1 | - | 499 | ||||
| U21661.1 | - | 3 | 457 | |||

Mtpn
Genomic Map