siRNA Details
| Gene Symbol: Gja3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024376.2 | NP_077352.1 | 2 | 880 | ||
| XM_008770777.1 | XP_008768999.1 | 2 | 728 | |||
| GenBank mRNA | X57970.1 | - | 2 | 882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gja3| Gene Symbol: Gja3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024376.2 | NP_077352.1 | 2 | 880 | ||
| XM_008770777.1 | XP_008768999.1 | 2 | 728 | |||
| GenBank mRNA | X57970.1 | - | 2 | 882 | ||