siRNA Details
| Gene Symbol: Gk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024381.2 | NP_077357.2 | 13 | 1102 | ||
| XM_006256987.2 | XP_006257049.1 | 14 | 1154 | |||
| XM_006256988.2 | XP_006257050.1 | 13 | 1136 | |||
| XM_008773274.1 | XP_008771496.1 | 15 | 1219 | |||
| XM_008773275.1 | XP_008771497.1 | 14 | 1201 | |||
| XM_008773276.1 | XP_008771498.1 | 15 | 1219 | |||
| XM_008773278.1 | XP_008771500.1 | 15 | 1218 | |||
| GenBank mRNA | CO564445.1 | - | 4 | 235 | ||
| D16102.1 | - | 13 | 1105 | |||

Gk
Genomic Map