siRNA Details
| Gene Symbol: Hmox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277073.1 | NP_001264002.1 | 4 | 451 | ||
| NM_024387.2 | NP_077363.1 | 4 | 495 | |||
| XM_006245827.2 | XP_006245889.1 | 4 | 673 | |||
| XM_008767500.1 | XP_008765722.1 | 3 | 1402 | |||
| GenBank mRNA | BC062061.1 | - | 4 | 410 | 5599345 | |
| FM134576.1 | - | 4 | 388 | |||
| J05405.1 | - | 4 | 487 | |||

Hmox2
Genomic Map