siRNA Details
| Gene Symbol: Hsd17b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024392.2 | NP_077368.2 | 16 | 1407 | ||
| GenBank mRNA | BC072472.1 | - | 16 | 1397 | 7100430 | |
| S83279.1 | - | 16 | 1379 | |||
| U37486.1 | - | 16 | 1407 | |||
| X94978.1 | - | 16 | 1382 | |||

Hsd17b4
Genomic Map