siRNA Details
| Gene Symbol: Hsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024393.1 | NP_077369.1 | 4 | 584 | ||
| XM_006241888.2 | XP_006241950.1 | 4 | 573 | |||
| XM_006241889.2 | XP_006241951.1 | 4 | 574 | |||
| XM_006241890.2 | XP_006241952.1 | 4 | 573 | |||
| XM_006241891.2 | XP_006241953.1 | 4 | 572 | |||
| GenBank mRNA | X83094.1 | - | 2 | 156 | ||

Hsf1
Genomic Map