siRNA Details
| Gene Symbol: Adamts1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024400.2 | NP_077376.2 | 9 | 2775 | ||
| GenBank mRNA | AF149118.1 | - | 9 | 2788 | ||
| AF304446.1 | - | 9 | 2346 | |||
| BC080237.1 | - | 9 | 2775 | 7100443 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts1| Gene Symbol: Adamts1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024400.2 | NP_077376.2 | 9 | 2775 | ||
| GenBank mRNA | AF149118.1 | - | 9 | 2788 | ||
| AF304446.1 | - | 9 | 2346 | |||
| BC080237.1 | - | 9 | 2775 | 7100443 | ||