siRNA Details
| Gene Symbol: Zfp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133290.3 | NP_579824.2 | 2 | 343 | ||
| GenBank mRNA | BC060308.1 | - | 2 | 347 | 6922188 | |
| X63369.1 | - | 2 | 328 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp36| Gene Symbol: Zfp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133290.3 | NP_579824.2 | 2 | 343 | ||
| GenBank mRNA | BC060308.1 | - | 2 | 347 | 6922188 | |
| X63369.1 | - | 2 | 328 | |||