siRNA Details
| Gene Symbol: Igfals | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053329.2 | NP_445781.2 | 2 | 1555 | ||
| GenBank mRNA | S46785.1 | - | 2 | 1555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfals| Gene Symbol: Igfals | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053329.2 | NP_445781.2 | 2 | 1555 | ||
| GenBank mRNA | S46785.1 | - | 2 | 1555 | ||