siRNA Details
| Gene Symbol: Fabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053365.1 | NP_445817.1 | 2 | 252 | ||
| GenBank mRNA | AF144756.1 | - | 2 | 252 | ||
| BC084721.1 | - | 2 | 240 | 7310595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp4| Gene Symbol: Fabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053365.1 | NP_445817.1 | 2 | 252 | ||
| GenBank mRNA | AF144756.1 | - | 2 | 252 | ||
| BC084721.1 | - | 2 | 240 | 7310595 | ||