siRNA Details
Gene Symbol: Timm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032618.1 | NP_116007.1 | 3 | 459 | ||
GenBank mRNA | AF223951.1 | - | 3 | 443 | ||
FM107575.1 | - | 3 | 459 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Timm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032618.1 | NP_116007.1 | 3 | 459 | ||
GenBank mRNA | AF223951.1 | - | 3 | 443 | ||
FM107575.1 | - | 3 | 459 |