siRNA Details
| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030585.1 | NP_085074.1 | 5 | 609 | ||
| GenBank mRNA | BC089795.1 | - | 5 | 595 | 7375879 | |
| U34932.1 | - | 5 | 609 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rabep2| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030585.1 | NP_085074.1 | 5 | 609 | ||
| GenBank mRNA | BC089795.1 | - | 5 | 595 | 7375879 | |
| U34932.1 | - | 5 | 609 | |||