siRNA Details
| Gene Symbol: Cubn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053332.2 | NP_445784.1 | 31, 32 | 4689 | ||
| GenBank mRNA | AF022247.1 | - | 31, 32 | 4689 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cubn| Gene Symbol: Cubn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053332.2 | NP_445784.1 | 31, 32 | 4689 | ||
| GenBank mRNA | AF022247.1 | - | 31, 32 | 4689 | ||